Learn More
PTP4A1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PTP4A1.
Supplier: Abnova Corporation PAB6077
Description
The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs), which contains a PTP domain and a characteristic C-terminal prenylation motif. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This tyrosine phosphatase is a nuclear protein, but may primarily associate with plasma membrane. The surface membrane association of this protein depends on its C-terminal prenylation. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which implicated its role in the tumorigenesis. Studies in rat suggested that this gene may be an immediate-early gene in mitogen-stimulated cells. [provided by RefSeq
Sequence: KDSNGHRNNCCIQSpecifications
PTP4A1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
PTP4A1 | |
PTP4A1 | |
A synthetic peptide corresponding to human PTP4A1. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PTP4A1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
DKFZp779M0721/HH72/PRL-1/PRL1/PTP(CAAX1)/PTPCAAX1 | |
Goat | |
Antigen affinity purification | |
RUO | |
7803 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only