Learn More
Goat anti-Rabbit IgG, Secondary Antibody Millipore™ (Chemicon™)
Manufacturer:  Millipore Sigma AP307P/DEL
 View more versions of this product
Catalog No. 50-174-508
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Goat anti-Rabbit IgG
Affinity Purified
IgG
RUO
Secondary
Rabbit
HRP
Goat
1mL
Enzyme
Polyclonal