Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Goat anti-Rabbit IgG, Whole Molecule, Affinity Gel

goat affinity purified antibody to rabbit IgG (whole molecule) covalently coupled to 2mL of Sepharose-4B, and packed in a polystyrene chromatography column

Manufacturer:  MP Biomedicals™ 0855696

Catalog No. MP0855696


Add to cart

Description

Description

Host Species: Goat
Species Reactivity: Rb

Antibody Purification

Specifications

Specifications

2mL
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit