Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG Fc Secondary Antibody, R-Phycoerythrin (Pre-adsorbed), Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP261920PE

Catalog No. NBP261920PE


Add to Cart

Description

Description

IgG Fc Polyclonal antibody specifically detects IgG Fc in Rat samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

IgG Fc
Polyclonal
R-PE
PBS containing 0.2% BSA with 0.09% Sodium Azide
Rat IgG-Fc Fragment
1 mL
Secondary
Rat
IgG
Flow Cytometry, Immunofluorescence
0.5 mg/mL
Flow Cytometry 1:50 - 1:200, Immunofluorescence 1:50 - 1:200
Goat
Affinity Purified
Adaptive Immunity
Antiserum was solid phase adsorbed to ensure class specificity. Antiserum was cross adsorbed using human and mouse immunosorbents to remove cross reactive antibodies. The antibody to rat IgG was isolated by affinity chromatography using antigen coupled to agarose beads. F(ab')2 fragments were generated using a pepsin digestion. Fc fragments and whole IgG molecules have been removed. Fragments were conjugated to R-phycoerythrin (RPE). Antibody concentration was determined by extinction coefficient: absorbance at 280 nm of 1.4 equals 1.0 mg of IgG. By immunoelectrophoresis and ELISA this antibody reacts specifically with rat IgG. Cross reactivity with IgA and IgM is negligible. No antibody was detected against non-immunoglobulin serum proteins. Less than 1% cross reactivity to human and mouse IgG was detected. This antibody may cross react with IgG from other species.
Store at 4C. Do not freeze.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit