Learn More
RNF103, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of RNF103.
Supplier: Abnova Corporation PAB6394
Description
The protein encoded by this gene contains a RING-H2 finger, a motif known to be involved in protein-protein and protein-DNA interactions. This gene is highly expressed in normal cerebellum, but not in the cerebral cortex. The expression of the rat counterpart in the frontal cortex and hippocampus was shown to be induced by elctroconvulsive treatment (ECT) as well as chronic antidepressant treatment, suggesting that this gene may be a molecular target for ECT and antidepressants. [provided by RefSeq
Sequence: C-YAQHQPLSNDVPSSpecifications
RNF103 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
RNF103 | |
RNF103 | |
A synthetic peptide corresponding to human RNF103. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of RNF103. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
KF1/MGC102815/MGC41857/ZFP103/hkf-1 | |
Goat | |
Antigen affinity purification | |
RUO | |
7844 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only