Learn More
SCARB2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SCARB2.
Supplier: Abnova Corporation PAB13673
Description
The protein encoded by this gene is a type III glycoprotein that is located primarily in limiting membranes of lysosomes and endosomes. Studies of the similar protein in mice and rat suggested that this protein may participate in membrane transportation and the reorganization of endosomal/lysosomal compartment. Deficiency of the similar protein in mice was reported to impair cell membrane transport processes and cause pelvic junction obstruction, deafness, and peripheral neuropathy. [provided by RefSeq
Sequence: C-NKANIQFGDNGTTISSpecifications
SCARB2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:4000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
SCARB2 | |
SCARB2 | |
A synthetic peptide corresponding to amino acids at internal region of human SCARB2. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SCARB2. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
AMRF/CD36L2/HLGP85/LIMPII/SR-BII | |
Goat | |
Antigen affinity purification | |
RUO | |
950 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only