Learn More
SLC26A6 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SLC26A6.
Supplier: Abnova Corporation PAB27544
Description
This gene belongs to the solute carrier 26 family, whose members encode anion transporter proteins. This particular family member encodes a protein involved in transporting chloride, oxalate, sulfate and bicarbonate. Several alternatively spliced transcript variants of this gene, encoding distinct isoforms, have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq
Sequence: SAPRTHQWRTSpecifications
SLC26A6 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (0.1-0.3mg/ml) Immunohistochemistry (0.1-0.3mg/ml) The optimal working dilution should be determined by the end user. | |
SLC26A6 | |
SLC26A6 | |
A synthetic peptide corresponding to internal region of human SLC26A6. | |
100 μg | |
Primary | |
Human |
Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SLC26A6. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
DKFZp586E1422 | |
Goat | |
Affinity Purified | |
RUO | |
65010 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only