Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SMAD4.
Supplier: Abnova Corporation PAB6049
Description
Sequence: C-HTMPIADPQPLDSpecifications
SMAD4 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
SMAD4 | |
SMAD4 | |
A synthetic peptide corresponding to human SMAD4. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SMAD4. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
DPC4/JIP/MADH4 | |
Goat | |
Antigen affinity purification | |
RUO | |
4089 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only