Learn More
SNCA Goat anti-Dog, Human, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SNCA.
Supplier: Abnova Corporation PAB27570
Description
Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. Four alternatively spliced transcripts encoding two different isoforms have been identified for this gene. [provided by RefSeq
Sequence: ATGFVKKDQLGKSpecifications
SNCA | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (0.1-0.3ug/ml) The optimal working dilution should be determined by the end user. | |
SNCA | |
SNCA | |
A synthetic peptide corresponding to internal region of human SNCA. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Porcine, Rat |
Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of SNCA. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
MGC110988/NACP/PARK1/PARK4/PD1 | |
Goat | |
Affinity Purified | |
RUO | |
6622 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only