Learn More
SOD2 Goat anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of SOD2.
Supplier: Abnova Corporation PAB19634
Description
This gene is a member of the iron/manganese superoxide dismutase family. It encodes a mitochondrial protein that forms a homotetramer and binds one manganese ion per subunit. This protein binds to the superoxide byproducts of oxidative phosphorylation and converts them to hydrogen peroxide and diatomic oxygen. Mutations in this gene have been associated with idiopathic cardiomyopathy (IDC), premature aging, sporadic motor neuron disease, and cancer. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: C-EAIKRDFGSFDKSpecifications
SOD2 | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of SOD2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
IPO-B/MNSOD/Mn-SOD | |
Goat | |
Antigen affinity purification | |
RUO | |
6648 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (0.01-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
SOD2 | |
SOD2 | |
A synthetic peptide corresponding to amino acids 119-130 at internal region of human SOD2. | |
100 μg | |
Primary | |
The immunizing peptide represents the acetylation site including K122 according to isoform A. This antibody is expected to recognize both reported isoforms (NP_000627.2; NP_001019637.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only