Learn More
STAT4 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of STAT4.
Manufacturer: Abnova Corporation PAB19011
Description
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein is essential for mediating responses to IL12 in lymphocytes, and regulating the differentiation of T helper cells. [provided by RefSeq
Sequence: C-DHSESGEVRFHSVEPSpecifications
STAT4 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.2-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
STAT4 | |
STAT4 | |
A synthetic peptide corresponding to amino acids 613-627 at internal region of human STAT4. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of STAT4. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
SLEB11 | |
Goat | |
Antigen affinity purification | |
RUO | |
6775 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only