Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFBR1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of TGFBR1.

Manufacturer:  Abnova Corporation PAB19758

Catalog No. 89-121-285


Add to cart

Description

Description

The protein encoded by this gene forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: TDGLCFVSVTETTD
Specifications

Specifications

TGFBR1
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of TGFBR1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Liquid
TGFBR1
A synthetic peptide correspoding to amino acids 50-63 at internal region of human TGFBR1.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_004603.1; NP_001124388.1). The immunizing peptide represents part of a potential extracellular domain.
ELISA, Western Blot
Unconjugated
ELISA (1:64000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
TGFBR1
AAT5|ACVRLK4|ALK-5|ALK5|LDS1A|LDS2A|SKR4|TGFR-1
Goat
Antigen affinity purification
RUO
Primary
7046
Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only