Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGM2 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of TGM2.

Supplier:  Abnova Corporation PAB7291

Catalog No. 89-133-119


Add to Cart

Description

Description

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, the encoded protein is the autoantigen implicated in celiac disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-NFESDKLKAVKGFR
Specifications

Specifications

TGM2
Polyclonal
Unconjugated
ELISA (1:128000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user.
TGM2
TGM2
A synthetic peptide corresponding to human TGM2.
100 μg
Primary
This antibody is expected to recognize isoform a (NP_004604.2) only.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of TGM2.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
G-ALPHA-h/GNAH/TG2/TGC
Goat
Antigen affinity purification
RUO
7052
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only