Learn More
TRIM8, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TRIM8.
Supplier: Abnova Corporation PAB6352
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes. [provided by RefSeq
Sequence: C-YGQPSTKHYVTSSpecifications
TRIM8 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user. | |
TRIM8 | |
TRIM8 | |
A synthetic peptide corresponding to C-terminus of human TRIM8. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of TRIM8. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
GERP/RNF27 | |
Goat | |
Antigen affinity purification | |
RUO | |
81603 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only