Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC7, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of TRPC7.
Manufacturer: Abnova Corporation PAB17030
Description
Sequence: KLCKSKAKSCENDSpecifications
TRPC7 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
TRPC7 | |
TRPC7 | |
A synthetic peptide corresponding to internal region of human TRPC7. | |
100 μg | |
Primary | |
Human |
ELISA, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of TRPC7. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
TRP7 | |
Goat | |
Antigen affinity purification | |
RUO | |
57113 | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only