Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBR2, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of UBR2.

Manufacturer:  Abnova Corporation PAB17264

Catalog No. 89-118-929


Add to cart

Description

Description

Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM

Sequence: SCPKSGGDKSRAPT
Specifications

Specifications

UBR2
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of UBR2.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Liquid
UBR2
A synthetic peptide corresponding to amino acids at internal region of human UBR2.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
UBR2
C6orf133|DKFZp686C08114|KIAA0349|MGC71112|RP3-392M17.3|bA49A4.1|dJ242G1.1|dJ392M17.3
Goat
Antigen affinity purification
RUO
Primary
23304
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only