Learn More
UCP1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of UCP1.
Supplier: Abnova Corporation PAB6905
Description
Mitochondrial uncoupling proteins (UCP) are members of the family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat. [provided by RefSeq
Sequence: C-EQLKRELSKSRQSpecifications
UCP1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (0.1-1 ug/mL) The optimal working dilution should be determined by the end user. | |
UCP1 | |
UCP1 | |
A synthetic peptide corresponding to human UCP1. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of UCP1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
SLC25A7/UCP | |
Goat | |
Antigen affinity purification | |
RUO | |
7350 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only