Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Antiserum to Sheep Serum (Forensic), MP Biomedicals™

Goat Antiserum to Sheep Serum (Forensic)

Manufacturer:  MP Biomedicals, Inc 0856006

Catalog No. MP0856006


Add to cart

Description

Description

  • Product is the lyophilized powder of goat antiserum to sheep serum and buffer salts.
  • Whole serum polyclonal antibody is used for immunostaining of cell suspensions, acetone-fixed frozen tissue sections, and formalin-fixed, paraffin-embedded tissue sections.
  • This product is intended for use as a precipitating antibody.
  • It gives sharp precipitin lines in immunoelectrophoresis, capillary tube, and immunodiffusion techniques.
Specifications

Specifications

Immunohistochemistry
Unconjugated
Lyophilized PBS with 0.05% Sodium Azide
Goat
RUO
Primary
Reconstitute product with 2.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material. If a small amount of precipitate is present, or if preparation
Total Protein: 62.6mg/mL
Product is the lyophilized powder ofgoat antiserum to sheep serum and buffer salts.
Lyophilized
2mL
Store product at 2-8°C. Store reconstituted material no longer than 4 weeks at 2-8°C. For long-term storage, aliquot and freeze reconstituted product at -20°C or lower.
Polyclonal
Sheep
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit