Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat IgG ELISA Kit (Colorimetric)

Useful in study of immunity

$509.50

Specifications

Species Goat
Sensitivity 0.134μg/mL
Sample Type Serum, plasma
Research Discipline Immunology
Synonym Immunoglobulin heavy constant gamma 1 (G1m marker)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600611KT View Documents Novus Biologicals™
NBP2600611KIT
1 Kit Each for $509.50
Add to cart
 
Description

Description

The Goat IgG ELISA Kit (Colorimetric) measures goat IgG in serum, plasma.
Specifications

Specifications

Goat
Serum, plasma
Immunoglobulin heavy constant gamma 1 (G1m marker)
IgG
ELISA Kit (Colorimetric)
50 to 100μL
0.134μg/mL
Immunology
Store unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
0.146 to 37.5μg/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit