Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG, F(ab)2, Agarose Immobilized, Isotype control, Novus Biologicals™

Goat Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197191

Catalog No. NBP197191


Add to cart

Description

Description

IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Goat samples. It is validated for Immunoprecipitation, SDS-Page.
Specifications

Specifications

IgG F(ab)2
Agarose
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
Goat
Multi-step
RUO
Primary
Goat
Immunoprecipitation, SDS-Page
Immunoprecipitation 1:10 - 1:500, SDS-Page 1:100 - 1:2000
Conjugated
IgG2
5 mg
Store at 4C. Do not freeze.
This product is a purified Goat IgG isolated from normal Goat Serum digested by the enzyme pepsin and purified by chromatography coupled to activated agarose. Sufficient antibody capacity is provided to bind a minimum of 5.0 mg of anti-Goat IgG.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit