Learn More
Goat IgG, F(ab)2, Fluorescein, Isotype control, Novus Biologicals™
Goat Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196838
Catalog No. NBP196838
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IgG F(ab)2
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Goat IgG, anti-Goat IgG F(ab)2 and anti-Goat Serum. No reaction was observed against anti-Goat IgG F(ab)2 or anti-Pepsin.
Goat
SDS-Page
Fluorescein
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Thimerosal
Goat
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.