Learn More
Goat IgG, F(ab)2, HRP, Isotype control, Novus Biologicals™
Goat Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197136
Catalog No. NBP197136
Add to cart
Description
Description
IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Goat samples. It is validated for SDS-Page.
Specifications
Specifications
IgG F(ab)2
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat IgG, anti-Goat IgG F(ab)2 and anti-Goat Serum. No reaction was observed against anti-Goat IgG F(c) or anti-Pepsin.
Goat
SDS-Page
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Gentamicin Sulfate
Goat
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.