Learn More
Goat IgG, Fc, Fluorescein, Isotype control, Novus Biologicals™
Goat Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196839
Catalog No. NBP196839
Add to cart
Description
Description
IgG Fc antibody specifically detects IgG Fc in Goat samples. It is validated for SDS-Page.
Specifications
Specifications
IgG Fc
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Goat IgG, anti-Goat IgG F(c) and anti-Goat Serum. No reaction was observed against anti-Goat IgG F(ab)2 or anti- Papain.
Goat
SDS-Page
Fluorescein
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
Goat
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.