Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG, H/L Chains, Agarose Immobilized, Isotype control, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197259

Catalog No. NBP197259


Add to cart

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Immunoprecipitation.
Specifications

Specifications

IgG (H+L)
Agarose
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Rabbit
IgG
20 mg
Store at 4C in the dark.
Polyclonal
Goat
Immunoprecipitation
Immunoprecipitation
Conjugated
Goat IgG whole molecule
IgG purified
RUO
Secondary
Anti-Goat IgG [H&L] (Rabbit) Antibody Agarose Conjugated is an IgG fraction antibody coupled to activated agarose. Sufficient antibody capacity is provided to bind a minimum of 5 mg of pure Goat IgG.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit