Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat Insulin ELISA Kit (Colorimetric)

Colorimetric ELISA Kit is useful in study of Apolipoproteins.

$702.50

Specifications

Species Goat
Sensitivity 2 uIU/mL
Sample Type Serum, plasma, tissue homogenates
Research Discipline Lipid and Metabolism
Synonym IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600741KT View Documents Novus Biologicals™
NBP2600741KIT
1 Kit Each for $702.50
Add to cart
 
Description

Description

The Goat Insulin ELISA Kit (Colorimetric) measures goat Insulin in serum, plasma, tissue homogenates.
Specifications

Specifications

Goat
Serum, plasma, tissue homogenates
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Insulin
3630
ELISA Kit (Colorimetric)
2 uIU/mL
Lipid and Metabolism
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
INS
8 to 100uIU/ml

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit