Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat Thyroxine ELISA Kit (Colorimetric)

Colorimetric ELISA Kit for study of thyroid function.

$602.00

Specifications

Sensitivity 20ng/mL
Sample Type Plasma, Serum, Tissue Homogenates
Research Discipline Signal Transduction
Synonym T4
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601571KT View Documents Novus Biologicals™
NBP2601571KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Goat Thyroxine ELISA Kit (Colorimetric) measures goat Thyroxine in serum, plasma, tissue homogenates.
Specifications

Specifications

20ng/mL
Signal Transduction
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
20 to 320ng/mL
Plasma, Serum, Tissue Homogenates
T4
Thyroxine
ELISA Kit (Colorimetric)

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit