Learn More
Cole-Parmer GoatThroat GT200-C/STANDOFF Pump With Epdm Seal, 4in Standoff
Manufacturer:  Cole-Parmer MFLX7565012
Encompass_Preferred
The GoatThroat pumps are designed to handle a wide range of liquids—wetted parts are 100% nonreactive, food-grade polypropylene. Choose nitrile seals for petroleum-based products including oil, diesel fuel, and glycerin. The EPDM seals are ideal for general-purpose chemicals including some acids, acetates, alcohols, and phosphates. Santoprene seals are ideal for acetic acid, acetone, and hydrocyanic acid among others. Viton seals were developed for more aggressive chemicals including bromine, sulfuric acid, ethylene, nitric acid, gasoline, and pesticides.
Catalog No. NC1820255