Learn More
Godax Labs™ NOCHROMIX™ Safety Kits
For safer handling of our NOCHROMIX solution
Manufacturer:  Godax Labs™ 21 003
 View more versions of this product
Catalog No. 02-883A
Add to cart
Description
Description
  • Kit comprises safety gloves and safety vent caps
  • No. of pair safety gloves and safety vent caps: 3
Specifications
Specifications
Storing Nochromix solution in the Sulfuric Acid Glass bottle, prevent pressure build-up by permitting gases to escape automatically as internal pressure increases
Safety Vent Cap