Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ HAND MEDIC™ Professional Skin Conditioner

Fragrance-free skin conditioner, specially formulated for industrial environments.

$83.50 - $113.70

Specifications

Color White
Product Type Skin Conditioner
For Use With (Application) Professional technicians, for maintaining natural protective barrier of skin
pH Range 3.5 to 5
View More Specs

Products 2
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Packaging Quantity Size (Metric) Price Quantity & Availability  
19-043-738
View Documents
GOJO™
814212
2 oz. Squeeze Bottle 12 Case 58.14mL
Case of 12 for $83.50
Add to cart
 
19-043-739
View Documents
GOJO™
814506
8 oz. Pump Bottle 6 Case 236mL
Case of 6 for $113.70
Add to cart
 
Description

Description

 

  • Helps prevent dry, cracked skin by maintaining skin's moisture level
  • Absorbs quickly with no greasy after-feel
  • Silicone and fragrance free
  • Dermatologist tested
  • Latex glove compatible
  • NSF E4 Registered

 

Specifications

Specifications

White
Skin Conditioner
Professional technicians, for maintaining natural protective barrier of skin
3.5 to 5
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit