GOJO Hand Medic Skin Conditioner

Manufacturer: GOJO INDUSTRIES INC 814506

 View more versions of this product

Catalog No. 19-043-739

  Check Availability
Add to Cart

    Specifications

    Composition Glycerin < 3%, Aluminum Starch Octenylsuccinate < 3%
    For Use With (Application) Professional technicians, for maintaining natural protective barrier of skin
    Product Type Skin Conditioner
    Size (English) 8 oz.
    Size (Metric) 236mL
    Packaging Bottle
    Quantity 6 Case
    Color White
    pH Range 3.5 to 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit