GOJO™ MULTI GREEN™ Hand Cleaners

Manufacturer: GOJO INDUSTRIES INC 917212

 View more versions of this product

Catalog No. 19-088286

  Check Availability
Add to Cart

    Specifications

    Color Green
    For Use With (Equipment) Bag-in-Box system
    Size (English) 0.21 gal.
    Size (Metric) 800mL
    Quantity 12 Case
    Composition Laureth-7
    For Use With (Application) For removing light to medium Industrial soils and Dirt
    Fragrance Citrus
    Product Type Multi Green™ Hand Cleaner
    pH Range 6 to 9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit