Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gold AA Standard, 1mL = 1mg Au (1,000ppm Au), Ricca Chemical
Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass amber
Specifications
Concentration or Composition Notes | 1,000ppm Au |
CAS Min % | 93.0,4.13,0.1 |
CAS Max % | 97.0,4.3,0.1 |
Physical Form | Liquid |
Packaging | Glass amber |
Equivalence | 1 mL = 1 mg Au |
Color | Undesignated |
Quantity | 100mL |
Chemical Name or Material | Gold AA Standard |