Alfa Aesar™ Gold Beryllium pellet, 2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface

Manufacturer: Alfa Aesar™ 45608KU

Quantity 5 each

 View more versions of this product

Catalog No. AA45608KU

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00003436
    Chemical Name or Material Gold Beryllium pellet, 2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface

    Specifications

    Physical Form Pellet
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00003436
    Weight ≈0.554g/pellet
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Quantity 5 each
    Avg. Mol. Wt. or Mol. Wt. Range Au:Be; 99:1 wt%
    Health Hazard 1 Danger
    Chemical Name or Material Gold Beryllium pellet, 2.00mm (0.08 in.) dia. x 10mm (0.39 in.) length, slightly pitted surface

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit