Alfa Aesar™ Gold flake, APS 1.5-3.0 micron, 99.96% (metals basis)

Manufacturer: Alfa Aesar™ 4215106

Quantity 5g

 View more versions of this product

Catalog No. AA4215106

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-57-5
    MDL Number MFCD00003436
    Chemical Name or Material Gold flake

    Specifications

    Particle Size APS 1.5 to 3.0 Micron
    TSCA TSCA
    Physical Form Flakes
    MDL Number MFCD00003436
    CAS 7440-57-5
    Quantity 5g
    Chemical Name or Material Gold flake
    Assay 99.96%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit