Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Gauze, 100 mesh woven from 0.064mm (0.0025 in.) dia. Wire, 99.99% (Metals basis), Alfa Aesar™

Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in.

Manufacturer:  Alfa Aesar 40931FY

Quantity 50 x 100mm

 View more versions of this product

Catalog No. AA40931FY


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-57-5
99.99% (Metals basis)
Gold
Gauze
MFCD00003436

Specifications

50 x 100mm
Gold colored gauze
99.99% (Metals basis)
Odorless
Gold
7440-57-5
Gauze
MFCD00003436
Gold colored
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit