Alfa Aesar™ Gold Germanium slug, 6.35mm (0.25in)dia x 6.35mm (0.25 in.) length, Premion™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 14173KG

Quantity 1 piece

 View more versions of this product

Catalog No. AA14173KG

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091706
    Chemical Name or Material Gold Germanium slug, 6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
    Grade Premion™

    Specifications

    Physical Form Slug
    MDL Number MFCD02091706
    Weight ≈2.9g/slug
    TSCA TSCA
    Odor Odorless
    Quantity 1 piece
    Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
    Chemical Name or Material Gold Germanium slug, 6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
    Assay 99.999%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit