Alfa Aesar™ Gold(I) chloride, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4336506

Quantity 5g

 View more versions of this product

Catalog No. AA4336506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-29-8
    Molecular Formula AuCl
    Formula Weight 232.42
    MDL Number MFCD00046175
    UN Number UN3260
    Merck Index 14,4515
    Chemical Name or Material Gold (I) chloride

    Specifications

    Formula Weight 232.42
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 10294-29-8
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Sensitivity Moisture Sensitive
    Molecular Formula AuCl
    MDL Number MFCD00046175
    Merck Index 14,4515
    Melting Point 289°C (dec.)
    Quantity 5g
    Odor Odorless
    UN Number UN3260
    Health Hazard 1 Danger
    Density 7.57g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    Physical Form Powder
    Chemical Name or Material Gold (I) chloride
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit