Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(I) chloride, 99+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
10294-29-8
AuCl
232.42
MFCD00046175
FDWREHZXQUYJFJ-UHFFFAOYSA-M
gold i chloride,gold monochloride,gold chloride,aurous chloride,gold chloride trihydride,acmc-1bvpa,gold monochloride, acid,gold i chloride trace metals basis,gold i chloride, saj special grade
27366
CHEBI:30078
chlorogold
Cl[Au]
10294-29-8
AuCl
232.42
MFCD00046175
FDWREHZXQUYJFJ-UHFFFAOYSA-M
gold i chloride,gold monochloride,gold chloride,aurous chloride,gold chloride trihydride,acmc-1bvpa,gold monochloride, acid,gold i chloride trace metals basis,gold i chloride, saj special grade
27366
CHEBI:30078
chlorogold
Cl[Au]
Assay Percent Range 99+%
Linear Formula AuCl
Merck Index 15, 4550
Solubility Information Solubility in water: decomposes
Formula Weight 232.42
Physical Form Powder
Percent Purity 99+%
Elemental Analysis 99.0% min. (ex Au)
Packaging Glass bottle
Solubility Solubility in water: decomposes.
Color Yellow to Green
Melting Point 289.0°C
Quantity 5g
Chemical Name or Material Gold(I) chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May be corrosive to metals.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 233-655-8