Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(I) cyanide, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
506-65-0
CAuN
222.985
MFCD00003437
IZLAVFWQHMDDGK-UHFFFAOYSA-N
gold i cyanide,gold cyanide au cn,gold monocyanide,goldcarbonitrile,acmc-1cueu,gold i cyanide, premion r,gold i cyanide trace metals basis
68172
gold(1+);cyanide
[C-]#N.[Au+]
506-65-0
CAuN
222.985
MFCD00003437
IZLAVFWQHMDDGK-UHFFFAOYSA-N
gold i cyanide,gold cyanide au cn,gold monocyanide,goldcarbonitrile,acmc-1cueu,gold i cyanide, premion r,gold i cyanide trace metals basis
68172
gold(1+);cyanide
[C-]#N.[Au+]
Linear Formula AuCN
Merck Index 15, 4551
Formula Weight 222.98
Physical Form Powder
Gold (Au) 87.0 min.
Packaging Glass bottle
Solubility Solubility in water: negligible. Other solubilities: practically insoluble in alcohol, ether and, diluted acids, soluble in ammonia, solutions of nacn
Color Yellow
Quantity 250mg
Chemical Name or Material Gold(I) cyanide

GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger