Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(I) cyanide, Yellow powder, Premion™, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3203106

Quantity 5g

 View more versions of this product

Catalog No. AA3203106

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

506-65-0
99.99% (Metals basis)
222.98
Gold(I) cyanide
Powder
AuCN
MFCD00003437

Specifications

506-65-0
99.99% (Metals basis)
222.98
Almond-like
Yellow
Gold(I) cyanide
Powder
AuCN
MFCD00003437
Moisture sensitive
5g

Safety and Handling

SDS & Safety Documents

Keep container tightly closed. Do not empty into drains. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H301-H310-H330-H315-H318-H290-H400-H410: Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packing Group: II

EINECSNumber : 208-049-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit