Gold(I) cyanide, Yellow powder, Premion™, 99.99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3203101

Quantity 0.25g

 View more versions of this product

Catalog No. AA3203101

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 506-65-0
    Physical Form Powder
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula AuCN
    Formula Weight 222.98
    MDL Number MFCD00003437
    Chemical Name or Material Gold(I) cyanide

    Specifications

    Physical Form Powder
    Formula Weight 222.98
    Quantity 0.25g
    Color Yellow
    DOT Information DOT Class: 6.1, Packing Group: II
    Health Hazard 2 Keep container tightly closed. Do not empty into drains. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
    EINECS Number 208-049-1
    Odor Almond-like
    Sensitivity Moisture sensitive
    Assay Percent Range 99.99% (Metals basis)
    Molecular Formula AuCN
    MDL Number MFCD00003437
    CAS 506-65-0
    Health Hazard 1 H301-H310-H330-H315-H318-H290-H400-H410: Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Gold(I) cyanide

    Keep container tightly closed. Do not empty into drains. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H301-H310-H330-H315-H318-H290-H400-H410: Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit