Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold nanoparticles, 40nm, supplied in 0.1mM PBS, 95%, OD1, 530 nm absorption

Click to view available options
Quantity:
25mL
100mL
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Quantity 100mL
Physical Form Liquid
Percent Purity 95%
Particle Size 40nm
Color Pink to Red
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS, 95%, OD: 1, 530 nm absorption

storageNote1 : 2° to 8°C