Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold nanoparticles, 90nm, amine-functionalized, 3kDa PEGylated, OD50, 564nm absorption

Click to view available options
Quantity:
0.5mL
1mL
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
Quantity 0.5mL
Particle Size 90nm
Physical Form Liquid
Color Pink to Red
Chemical Name or Material Gold nanoparticles, amine-functionalized, 3kDa PEGylated, OD: 50, 564nm absorption

storageNote1 : 2° to 8°C