Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold nanoparticles, 90nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 564nm absorption

Click to view available options
Quantity:
25mL
100mL
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Quantity 100mL
Physical Form Liquid
Particle Size 90nm
Color Pink to Red
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 564nm absorption

storageNote1 : 2° to 8°C