Alfa Aesar™ Gold nanorods, 19-25nm dia. x 50-60nm length, dispersion in water, 650nm Absorption, positively charged

Manufacturer: Alfa Aesar™ 46331AC

Quantity 25mL

 View more versions of this product

Catalog No. AA46331AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-57-5
    MDL Number MFCD00003436
    Chemical Name or Material Gold nanorods, 19 to 25nm dia. x 50 to 60nm length, dispersion in water, 650nm Absorption, positively charged

    Specifications

    Physical Form Liquid
    MDL Number MFCD00003436
    Boiling Point >60°C (dec.)
    Quantity 25mL
    TSCA TSCA
    CAS 7440-57-5
    Odor Odorless
    Color Colorless
    Chemical Name or Material Gold nanorods, 19 to 25nm dia. x 50 to 60nm length, dispersion in water, 650nm Absorption, positively charged

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit