Alfa Aesar™ Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged

Manufacturer: Alfa Aesar™ 46923AC

Quantity 25mL

 View more versions of this product

Catalog No. AA46923AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-57-5
    MDL Number MFCD00003436
    Chemical Name or Material Gold nanorods, 19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged

    Specifications

    MDL Number MFCD00003436
    CAS 7440-57-5
    Color Colorless
    Quantity 25mL
    TSCA TSCA
    Physical Form Liquid
    Odor Odorless
    Boiling Point >60°C (dec.)
    Chemical Name or Material Gold nanorods, 19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit