Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold nanorods, 19-25nm dia. x 63-73nm length, dispersion in water, 700nm Absorption, negatively charged

Manufacturer:  Alfa Aesar™ 46923AA

Quantity 5mL

 View more versions of this product

Catalog No. AA46923AA

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-57-5
MFCD00003436
Gold nanorods, 19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged

Specifications

5mL
Liquid
Odorless
>60°C (dec.)
7440-57-5
MFCD00003436
Colorless
Gold nanorods, 19 to 25nm dia. x 63 to 73nm length, dispersion in water, 700nm Absorption, negatively charged

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit