Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold powder, spherical, <5 micron, 99.95% (metals basis)

Click to view available options
Quantity:
1g
5g
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
Quantity 1g
Physical Form Sperical Powder, S.A. 0.10-0.35m
Particle Size <5 Micron
Percent Purity 99.95%
Surface Area 0.10 to 0.35m2/g
Chemical Name or Material Gold powder
Assay (metals basis)

EINECSNumber : 231-165-9

TSCA : Yes

Recommended Storage : Ambient temperatures