Learn More
Thermo Scientific™ Gold Seal™ Rite-On™ and Ultra Frost™ Frosted Microscope Slides Available on GSA/VA Contract for Federal Government customers only.
Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass.
Manufacturer:  Thermo Scientific™ 3056002
 View more versions of this product
Catalog No. 12-518-107
  Check Availability
Add to cart
Specifications
Description
Label slides quickly and easily with Thermo Scientific™ Gold Seal™ Rite-On™ and  Ultra Frost™ Frosted Microscope slides, made from hard, colorless, noncorrosive glass. Choose from sandblast frosted end or white epoxy frosted end for easy marking. Each tab is impervious to virtually all common laboratory chemicals and reagents.
Description & Specifications
Specifications
Rite-On Ultra Frost Frosted Slide
Ground
75mm
25mm
90°
Packed 72 slides per box, 50 boxes per case
1mm
3 in.
1 in.
White Epoxy Ink Frosted End on Front Side with GOLD SEAL Logo Printed Key; Sandblast Frosted End on Back Side
3 x 1 in. (75 x 25mm)