Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold shot, 0.8-6mm (0.03-0.2 in.), Premion™, 99.999% (metals basis)

Click to view available options
Quantity:
1g
5g
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
7440-57-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
colloidal,powder,flake,leaf,gold, colloidal,burnish,shell,ci pigment metal 3,magnesium purple,c.i. pigment metal 3
23985
CHEBI:30050
gold
[Au]
Quantity 1g
Odor Odorless
Physical Form Shot
Percent Purity 99.999%
Grade Premion™
Chemical Name or Material Gold shot, 0.8 to 6mm (0.03 to 0.2 in.)
Assay (metals basis)

EINECSNumber : 231-165-9

RTECSNumber : MD5070000

TSCA : Yes

Recommended Storage : Ambient temperatures